OriGene
Manufacturer Code:CF813015
Catalog # 50-167-1046
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Chinese hamster |
Class | Monoclonal |
Type | Antibody |
Clone | OTI8C8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Hamster Monoclonal Antibody
For Research Use Only
Protein Aliases: B7 homolog 1; B7-H B7H1 PD-L1 PDCD1L1 PDCD1LG1 PDL1; B7-H1; CD274; CD274 antigen; PD-L1; PDCD1 ligand 1; Programmed cell death 1 ligand 1
Gene Aliases: B7-H; B7H1; CD274; PD-L1; PDCD1L1; PDCD1LG1; PDL1
UniProt ID: (Human) Q9NZQ7
Entrez Gene ID: (Human) 29126
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.