Invitrogen
Manufacturer Code:PA518337
Catalog # PIPA518337
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (CKKQSDTHLEET) corresponding to the C-terminus amino acids of CD274 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: B7 homolog 1; B7-H B7H1 PD-L1 PDCD1L1 PDCD1LG1 PDL1 B7 homolog 1 CD274 antigen PDCD1 ligand 1 programmed cell death 1 ligand 1 programmed death ligand 1; B7-H1; CD274; CD274 antigen; PD-L1; PDCD1 ligand 1; Programmed cell death 1 ligand 1; programmed death ligand 1
Gene Aliases: A530045L16Rik; B7-H; B7H1; CD274; PD-L1; PDCD1L1; PDCD1LG1; PDL1
UniProt ID: (Human) Q9NZQ7, (Mouse) Q9EP73
Entrez Gene ID: (Human) 29126, (Mouse) 60533
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
membrane-bound signaling molecule
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.