Invitrogen
Manufacturer Code:PA552285
Catalog # PIPA552285
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PCYT1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCT B; CCT-beta; Choline-phosphate cytidylyltransferase B; Choline-phosphate cytidylyltransferase B Phosphorylcholine transferase B CTP:phosphocholine cytidylyltransferase B CCT B CT B CCT-beta PCYT1B CCTB; CT B; CTP:phosphocholine cytidylyltransferase B; Phosphorylcholine transferase B
Gene Aliases: CCTB; CTB; PCYT1B
UniProt ID: (Human) Q9Y5K3
Entrez Gene ID: (Human) 9468
Molecular Function: nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.