Novus Biologicals
Manufacturer Code:NBP219734
Catalog # NBP219734
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human PCYT1B. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCT B; CCT B CCTB CCT-betaEC 2.7.7.15 choline-phosphate cytidylyltransferase B CT B CTB CTP:phosphocholine cytidylyltransferase B EC 2.7.7 phosphate cytidylyltransferase 1 choline beta phosphate cytidylyltransferase 1 choline beta isoform Phosphorylcholine transferase B; CCT-beta; Choline-phosphate cytidylyltransferase B; CT B; CTP:phosphocholine cytidylyltransferase B; Phosphorylcholine transferase B
Gene Aliases: CCTB; CTB; PCYT1B
UniProt ID: (Human) Q9Y5K3
Entrez Gene ID: (Human) 9468
Molecular Function:
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.