Proteintech
Manufacturer Code:137651AP150UL
Catalog # 50-172-8993
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PCYT1B Fusion Protein Ag4748 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCT B; CCT B CCT beta CCTB CT B CTB PCYT1B; CCT-beta; choline phosphate cytidylyltransferase 1 beta; Choline-phosphate cytidylyltransferase B; CT B; CTP:phosphocholine cytidylyltransferase B; phosphate cytidylyltransferase 1, choline, beta isoform; Phosphorylcholine transferase B
Gene Aliases: AW045697; CCT-beta; CCTB; Cctbeta; CTB; CTTbeta; PCYT1B
UniProt ID: (Human) Q9Y5K3, (Mouse) Q3UEW0, (Rat) Q9QZC4
Entrez Gene ID: (Human) 9468, (Mouse) 236899, (Rat) 286936
Molecular Function:
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.