Invitrogen
Manufacturer Code:MA538147
Catalog # PIMA538147
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2098 |
Immunogen | A synthesized peptide derived from human PCYT1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: CCT A; CCT A CCTA Cctalpha CCT-alpha choline phosphate cytidylyltransferase 1 alpha choline-phosphate cytidylyltransferase A CT CT A CTA CTalpha CTP CTP phosphocholine cytidylyl transferase CTP:phosphocholine cytidylyl transferase CTP:phosphocholine cytidylyltransferase A CTP:phosphocholine cytidylyltransferase alpha CTPCT Cttalpha PCYT1 Pcyt1a phosphate cytidylyltransferase 1 choline alpha phosphate cytidylyltransferase 1 choline alpha isoform phosphorylcholine transferase A SMDCRD; CCT-alpha; Choline-phosphate cytidylyltransferase A; CT A; CTP:phosphocholine cytidylyltransferase A; Phosphorylcholine transferase A
Gene Aliases: CCTA; CT; CTA; CTPCT; PCYT1; PCYT1A; SMDCRD
UniProt ID: (Human) P49585
Entrez Gene ID: (Human) 5130
Molecular Function: nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.