Novus Biologicals
Manufacturer Code:NBP297692R
Catalog # NB103067
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human PCYT1A fragment. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCT A; CCT A CCT-alpha choline-phosphate cytidylyltransferase A CT CT A CTA CTP:phosphocholine cytidylyltransferase A CTPCTphosphate cytidylyltransferase 1 choline alpha isoform EC 2.7.7 EC 2.7.7.15 PCYT1CCTA phosphate cytidylyltransferase 1 choline alpha Phosphorylcholine transferase A; CCT-alpha; Choline-phosphate cytidylyltransferase A; CT A; CTP:phosphocholine cytidylyltransferase A; Phosphorylcholine transferase A
Gene Aliases: CCTA; CT; CTA; CTPCT; PCYT1; PCYT1A; SMDCRD
UniProt ID: (Human) P49585
Entrez Gene ID: (Human) 5130
Molecular Function: nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.