Invitrogen
Manufacturer Code:PA514218
Catalog # PIPA514218
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 479-508 amino acids from the C-terminal region of human PCSK9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: convertase subtilisin/kexin type 9 preproprotein; NARC-1; neural apoptosis regulated convertase 1; Neural apoptosis-regulated convertase 1; PC9; Proprotein convertase 9; Proprotein convertase subtilisin/kexin type 9; Proprotein convertase subtilisin/kexin type 9 Proprotein convertase PC9 Subtilisin/kexin-like protease PC9 Neural apoptosis-regulated convertase 1 NARC-1 PSEC0052 NARC1 PCSK9; Subtilisin/kexin-like protease PC9
Gene Aliases: FH3; HCHOLA3; LDLCQ1; NARC-1; NARC1; PC9; PCSK9; PSEC0052
UniProt ID: (Human) Q8NBP7
Entrez Gene ID: (Human) 255738
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.