Invitrogen
Manufacturer Code:PA571918
Catalog # PIPA571918
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 659-688 amino acids from the C-terminal region of human PCSK6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Paired basic amino acid cleaving enzyme 4; paired basic amino acid cleaving system 4; Proprotein convertase subtilisin/kexin type 6; Proprotein convertase subtilisin/kexin type 6 3421- Paired basic amino acid cleaving enzyme 4 Subtilisin-like proprotein convertase 4 SPC4 Subtilisin/kexin-like protease PACE4 PCSK6 PACE4; SPC4; Subtilisin-like proprotein convertase 4; Subtilisin/kexin-like protease PACE4
Gene Aliases: PACE4; PCSK6; SPC4
UniProt ID: (Human) P29122
Entrez Gene ID: (Human) 5046
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.