Invitrogen
Manufacturer Code:PA568543
Catalog # PIPA568543
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human PCSK6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: kexin-like protease PC7A; PACE4 SPC4 paired basic amino acid cleaving enzyme 4 paired basic amino acid cleaving system 4 subtilisin-like proprotein convertase 4 subtilisin/kexin-like protease PACE4; Paired basic amino acid cleaving enzyme 4; paired basic amino acid cleaving system 4; Proprotein convertase subtilisin/kexin type 6; SPC4; Subtilisin - like endoprotease; Subtilisin-like proprotein convertase 4; Subtilisin/kexin-like protease PACE4
Gene Aliases: C86343; im:7140527; PACE4; PCSK6; SPC4
UniProt ID: (Human) P29122, (Rat) Q63415
Entrez Gene ID: (Dog) 488703, (Human) 5046, (Pig) 100524545, (Horse) 100057677, (Rabbit) 100347173, (Rat) 25507, (Mouse) 18553, (Zebrafish) 794305
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.