Abnova
Manufacturer Code:PAB30379
Catalog # 89-032-723
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to human PCSK6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PACE4; Paired basic amino acid cleaving enzyme 4; paired basic amino acid cleaving system 4; Proprotein convertase subtilisin/kexin type 6; SPC4; Subtilisin-like proprotein convertase 4; Subtilisin/kexin-like protease PACE4
Gene Aliases: PACE4; PCSK6; SPC4
UniProt ID: (Human) P29122
Entrez Gene ID: (Human) 5046
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.