Proteintech
Manufacturer Code:151061AP150UL
Catalog # 50-172-9983
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PCSK4 Fusion Protein Ag4007 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KEX2-like endoprotease 3; NEC 3; Neuroendocrine convertase 3; PC4; PC4 PCSK4 Proprotein convertase 4 SPC5; PCSK4; Prohormone convertase 3; Proprotein convertase 4; Proprotein convertase subtilisin/kexin type 4; testicular tissue protein Li 135
Gene Aliases: AI647044; L21221; NEC 3; Nec-3; NEC3; PC4; PCSK4; SPC5; UNQ2757/PRO6496
UniProt ID: (Human) Q6UW60, (Rat) Q78EH2, (Mouse) P29121
Entrez Gene ID: (Human) 54760, (Rat) 171085, (Mouse) 18551
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.