Invitrogen
Manufacturer Code:PA570441
Catalog # PIPA570441
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human PCSK2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KEX2-like endoprotease 2; NEC 2; Neuroendocrine convertase 2; Neuroendocrine convertase 2 NEC 2 Prohormone convertase 2 Proprotein convertase 2 PC2 KEX2-like endoprotease 2 NEC2 PCSK2; PC2; PC2-like protein; Prohormone convertase 2; propotein convetase subtilisin/kesin type 2; Proprotein convertase 2; proprotein convertase type 2
Gene Aliases: 6330411F23Rik; AI839700; NEC 2; NEC-2; NEC2; PC2; PCSK2; Phpp-2; Rpc2; SPC2
UniProt ID: (Human) P16519, (Bovine) Q9GLR0, (Rat) P28841, (Mouse) P21661
Entrez Gene ID: (Human) 5126, (Dog) 485757, (Horse) 100050755, (Bovine) 281968, (Rabbit) 100358339, (Rat) 25121, (Mouse) 18549, (Zebrafish) 100004460
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.