Invitrogen
Manufacturer Code:PA514595
Catalog # PIPA514595
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 87-116 amino acids from the N-terminal region of human PCSK2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KEX2-like endoprotease 2; NEC 2; Neuroendocrine convertase 2; Neuroendocrine convertase 2 NEC 2 Prohormone convertase 2 Proprotein convertase 2 PC2 KEX2-like endoprotease 2 NEC2 PCSK2; PC2; Prohormone convertase 2; Proprotein convertase 2
Gene Aliases: 6330411F23Rik; AI839700; NEC 2; NEC-2; NEC2; PC2; PCSK2; Phpp-2; SPC2
UniProt ID: (Human) P16519, (Mouse) P21661
Entrez Gene ID: (Human) 5126, (Mouse) 18549
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.