Invitrogen
Manufacturer Code:PA125306
Catalog # PIPA125306
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KEX2-like endoprotease 2; NEC 2; NEC2 PC2 SPC2; Neuroendocrine convertase 2; PC2; Prohormone convertase 2; Proprotein convertase 2
Gene Aliases: NEC 2; NEC-2; NEC2; PC2; PCSK2; Rpc2; SPC2
UniProt ID: (Human) P16519, (Rat) P28841
Entrez Gene ID: (Human) 5126, (Rat) 25121
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.