Invitrogen
Manufacturer Code:PA570424
Catalog # PIPA570424
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of rat PCSK1N |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: b-LEN; b-PEN-LEN; b-SAAS; basic protein IA-4; Big LEN; big PEN LEN; Big PEN-LEN; Big SAAS; granin-like neuroendocrine peptide; IA-4; KEP; l-LEN; l-SAAS; Little LEN; Little SAAS; N-proSAAS; pancreas protein 3; PEN; PEN-19; PEN-20; pro-SAAS; Proprotein convertase 1 inhibitor; Proprotein convertase subtilisin/kexin type 1 inhibitor; ProSAAS; protein serine alanine alanine serine; SAAS CT(1-49); SAAS CT(25-40)
Gene Aliases: AI848336; BigLEN; bLEN; KEP; lLEN; Pan3; PCSK1N; PEN; PEN19; PEN20; PROSAAS; SAAS; SAASCT; SCG8; SgVIII
UniProt ID: (Human) Q9UHG2, (Mouse) Q9QXV0, (Rat) Q9QXU9
Entrez Gene ID: (Human) 27344, (Bovine) 513603, (Dog) 491870, (Guinea pig) 100731521, (Mouse) 30052, (Rat) 246333
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.