Novus Biologicals
Manufacturer Code:NBP180797
Catalog # NBP180797
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:AETGAPRRFRRSVPRGEAAGAVQELARALAHLLEAERQERARAEAQEAEDQQARVLAQLLRVWGAPRNSDPALGLDDDPDAPAAQLARALLRARLDPAALAAQLVP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: b-LEN; b-PEN-LEN; b-SAAS; Big LEN; Big PEN-LEN; Big SAAS; granin-like neuroendocrine peptide; granin-like neuroendocrine peptide Proprotein convertase 1 inhibitor proprotein convertase subtilisin/kexin type 1 inhibitorPROSAAS proSAAS pro-SAAS SAAS; KEP; l-LEN; l-SAAS; Little LEN; Little SAAS; N-proSAAS; PEN; pro-SAAS; Proprotein convertase 1 inhibitor; Proprotein convertase subtilisin/kexin type 1 inhibitor; ProSAAS; SAAS CT(1-49); SAAS CT(25-40)
Gene Aliases: BigLEN; PCSK1N; PEN; PROSAAS; SAAS; SCG8; SgVIII
UniProt ID: (Human) Q9UHG2
Entrez Gene ID: (Human) 27344
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.