Invitrogen
Manufacturer Code:MA535027
Catalog # PIMA535027
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL5306 |
Immunogen | Recombinant Protein corresonding to Human PCP4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: brain specific polypeptide PEP19; brain specific polypeptide PEP19 brain-specific antigen PCP-4 Brain-specific polypeptide PEP-19 Calmodulin regulator protein PCP4 P16Rimb19 PCP4 Pcp-4 Pep19 PEP-19 PEPZ19 Purkinje cell protein 4; Brain-specific antigen PCP-4; Brain-specific polypeptide PEP-19; Calmodulin regulator protein PCP4; Purkinje cell protein 4
Gene Aliases: P16Rimb19; Pcp-4; PCP4; PEP-19; PEP19; PEPZ19
UniProt ID: (Human) P48539, (Mouse) P63054, (Rat) P63055
Entrez Gene ID: (Human) 5121, (Mouse) 18546, (Rat) 25510
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.