Novus Biologicals
Manufacturer Code:NBP261410
Catalog # NBP261410
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL5306 |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: GAGATNGKDKTSGENDGQKKVQEEFDIDMDAPETERAAVAIQSQFRKFQKKK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: brain specific polypeptide PEP19; brain specific polypeptide PEP19 10Brain-specific antigen PCP-4 Brain-specific polypeptide PEP-19 PEP19 PEP-19 Purkinje cell protein 4; brain-specific antigen PCP-4; Brain-specific polypeptide PEP-19; Calmodulin regulator protein PCP4; Purkinje cell protein 4
Gene Aliases: PCP4; PEP-19; PEP19
UniProt ID: (Human) P48539
Entrez Gene ID: (Human) 5121
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.