Proteintech
Manufacturer Code:240361AP150UL
Catalog # 50-173-4217
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PCNA Fusion Protein Ag7416 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cyclin; Cyclin PCNA; DNA polymerase delta auxiliary protein; PCNA; Proliferating cell nuclear antigen
Gene Aliases: ATLD2; PCNA; Pcna/cyclin; PCNAR
UniProt ID: (Human) P12004, (Rat) P04961, (Mouse) P17918
Entrez Gene ID: (Human) 5111, (Rat) 25737, (Mouse) 18538
Molecular Function:
DNA binding protein
DNA polymerase processivity factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.