Novus Biologicals
Manufacturer Code:NBP2854510.1ML
Catalog # NB033380
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human PCMD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C20orf36 chromosome 20 open reading frame 36 FLJ10883 KIAA0835 KIAA1050 protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 2 protein-L-isoaspartate O-methyltransferase domain-containing protein 2; Protein-L-isoaspartate O-methyltransferase domain-containing protein 2
Gene Aliases: C20orf36; PCMTD2
UniProt ID: (Human) Q9NV79
Entrez Gene ID: (Human) 55251
Molecular Function: methyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.