Novus Biologicals
Manufacturer Code:NBP154370
Catalog # NBP154370
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to PCMTD1(protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1) The peptide sequence was selected from the middle region of PCMTD1. Peptide sequence TGQNTWESKNILAVSFAPLVQPSKNDNGKPDSV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ10883 protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1 protein-L-isoaspartate O-methyltransferase domain-containing protein 1; Protein-L-isoaspartate O-methyltransferase domain-containing protein 1
Gene Aliases: PCMTD1
UniProt ID: (Human) Q96MG8
Entrez Gene ID: (Human) 115294
Molecular Function:
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.