Invitrogen
Manufacturer Code:MA524589
Catalog # PIMA524589
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL0206 |
Immunogen | Recombit protein corresponding to Human PCM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: hPCM-1; PCM-1; Pericentriolar material 1 protein; Pericentriolar material 1 protein PCM-1 hPCM-1 PCM1; pericentriolar material 1, PCM1
Gene Aliases: PCM1; PTC4; RET/PCM-1
UniProt ID: (Human) Q15154
Entrez Gene ID: (Human) 5108
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.