Novus Biologicals
Manufacturer Code:NBP275610
Catalog # NBP275610
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JB52-39 |
Immunogen | Recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: EC 4.1.1.32 PE PEPCK PEPCK2PEP carboxykinase PEPCK-M phosphoenolpyruvate carboxykinase [GTP] mitochondrial phosphoenolpyruvate carboxykinase 2 (mitochondrial) Phosphoenolpyruvate carboxylase phosphopyruvate carboxylase; PEP carboxykinase; PEPCK-M; phosphoenolpyruvate carboxykinase 2 (mitochondrial); Phosphoenolpyruvate carboxykinase [GTP], mitochondrial; phosphopyruvate carboxylase
Gene Aliases: PCK2; PEPCK; PEPCK-M; PEPCK2
UniProt ID: (Human) Q16822
Entrez Gene ID: (Human) 5106
Molecular Function:
decarboxylase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.