R&D Systems
Manufacturer Code:AF7316
Catalog # AF7316
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human PCK1 Met1-Ile88 Accession # P35558 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 4.1.1.32 MGC22652 PEPCK1 PEPCKC PEPCK-C PEPCK-CPEP carboxykinase phosphoenolpyruvate carboxykinase 1 (soluble) phosphoenolpyruvate carboxykinase cytosolic phosphoenolpyruvate carboxykinase cytosolic [GTP] Phosphoenolpyruvate carboxylase phosphopyruvate carboxylase; PEP carboxykinase; PEPCK-C; phosphoenolpyruvate carboxykinase 1 (soluble); phosphoenolpyruvate carboxykinase, cytosolic; Phosphoenolpyruvate carboxykinase, cytosolic [GTP]; phosphoenolpyruvate carboxylase; phosphopyruvate carboxylase; Serine-protein kinase PCK1
Gene Aliases: PCK1; PEPCK-C; PEPCK1; PEPCKC
UniProt ID: (Human) P35558
Entrez Gene ID: (Human) 5105
Molecular Function:
decarboxylase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.