Abnova
Manufacturer Code:PAB31373
Catalog # 89-998-362
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to human PCK1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC22652; PEP carboxykinase; PEPCK-C; PEPCK1; PEPCKC; phosphoenolpyruvate carboxykinase 1 (soluble); phosphoenolpyruvate carboxykinase, cytosolic; Phosphoenolpyruvate carboxykinase, cytosolic [GTP]; phosphoenolpyruvate carboxylase; phosphopyruvate carboxylase; Serine-protein kinase PCK1
Gene Aliases: PCK1; PEPCK-C; PEPCK1; PEPCKC
UniProt ID: (Human) P35558
Entrez Gene ID: (Human) 5105
Molecular Function: decarboxylase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.