Proteintech
Manufacturer Code:271031AP150UL
Catalog # 50-173-5564
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PCDH24 Fusion Protein Ag25840 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cadherin-related family member 2; CDHR2 PC LKC PCDH24 PCLKC protocadherin 24 Protocadherin LKC; PC-LKC; protocadherin 24; Protocadherin LKC; Protocadherin-24
Gene Aliases: CDHR2; Gm624; PC-LKC; PCDH24; PCLKC
UniProt ID: (Human) Q9BYE9, (Mouse) E9Q7P9
Entrez Gene ID: (Human) 54825, (Mouse) 268663
Molecular Function:
cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.