Proteintech
Manufacturer Code:665011IG150UL
Catalog # 50-173-6924
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1A9A5 |
Immunogen | PCCB Fusion Protein Ag1608 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: PCCase subunit beta; PCCase subunit beta PCCB; Propanoyl-CoA:carbon dioxide ligase subunit beta; propionyl CoA carboxylase, beta polypeptide; propionyl Coenzyme A carboxylase, beta polypeptide; propionyl-coA carboxylase B; Propionyl-CoA carboxylase beta chain, mitochondrial; propionyl-CoA carboxylase beta subunit
Gene Aliases: 1300012P06Rik; AI314687; PCCB; R74805
UniProt ID: (Pig) P79384, (Human) P05166, (Mouse) Q99MN9
Entrez Gene ID: (Pig) 100158147, (Human) 5096, (Mouse) 66904, (Rat) 24624
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.