Invitrogen
Manufacturer Code:PA576738
Catalog # PIPA576738
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human PCCB. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PCCase subunit beta; PCCase subunit betapccBPCCB&lowbarHUMANpccBC Complementation groupPropanoyl CoA:carbon dioxide ligase subunit betaPropanoyl-CoA:carbon dioxide ligase subunit betaPropionyl CoA carboxylase beta chain mitochondrialpropionyl Coenzyme A carboxylase beta polypeptidePropionyl-CoA carboxylase beta subunitPropionyl-CoA carboxylase beta chain mitochondrialR74805; Propanoyl-CoA:carbon dioxide ligase subunit beta; propionyl CoA carboxylase, beta polypeptide; propionyl Coenzyme A carboxylase, beta polypeptide; Propionyl-CoA carboxylase beta chain, mitochondrial
Gene Aliases: 1300012P06Rik; AI314687; PCCB; R74805
UniProt ID: (Human) P05166, (Mouse) Q99MN9
Entrez Gene ID: (Human) 5096, (Mouse) 66904
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.