Invitrogen
Manufacturer Code:PA530117
Catalog # PIPA530117
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 482 and 716 of Human PCCA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: pccA complementation group; PCCase alpha subunit; PCCase alpha subunit pccA complementation group propanoyl-CoA:carbon dioxide ligase alpha subunit propionyl Coenzyme A carboxylase alpha polypeptide propionyl-CoA carboxylase alpha chain mitochondrial; PCCase subunit alpha; propanoyl-CoA:carbon dioxide ligase alpha subunit; Propanoyl-CoA:carbon dioxide ligase subunit alpha; propionyl CoA carboxylase, alpha polypeptide; propionyl CoA-carboxylase alpha; Propionyl Coenzyme A carboxylase alpha polypeptide; propionyl Coenzyme A carboxylase, alpha polypeptide; Propionyl-CoA carboxylase alpha chain, mitochondrial; propionyl-coenzyme A carboxylase, alpha polypeptide
Gene Aliases: C79630; PCCA
UniProt ID: (Human) P05165, (Rat) P14882, (Mouse) Q91ZA3
Entrez Gene ID: (Human) 5095, (Rat) 687008, (Mouse) 110821
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.