Novus Biologicals
Manufacturer Code:NBP213736
Catalog # NBP213736
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: YPLRHKQADIRINGWAVECRVYAEDPYKSFGLPSIGRLSQYQEPLHLPGV RVDSGIQPGSDISIYYDPMISKLITYGSDRTEALKRMADALDNY |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 6.4.1 EC 6.4.1.3 pccA complementation group PCCase alpha subunit PCCase subunit alpha propanoyl-CoA:carbon dioxide ligase alpha subunit Propanoyl-CoA:carbon dioxide ligase subunit alpha propionyl CoA carboxylase alpha polypeptide propionyl Coenzyme A carboxylase alpha polypeptide propionyl-CoA carboxylase alpha chain mitochondrial; pccA complementation group; PCCase alpha subunit; PCCase subunit alpha; propanoyl-CoA:carbon dioxide ligase alpha subunit; Propanoyl-CoA:carbon dioxide ligase subunit alpha; propionyl CoA carboxylase, alpha polypeptide; propionyl Coenzyme A carboxylase, alpha polypeptide; Propionyl-CoA carboxylase alpha chain, mitochondrial
Gene Aliases: PCCA
UniProt ID: (Human) P05165
Entrez Gene ID: (Human) 5095
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.