Invitrogen
Manufacturer Code:PA5110076
Catalog # PIPA5110076
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-104 of human PCBD1 (NP_000272.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4-alpha-hydroxy-tetrahydropterin dehydratase; 4-alpha-hydroxy-tetrahydropterin dehydratase DCOH Dimerization cofactor of hepatocyte nuclear factor 1-alpha Pcbd PCBD1 PCD Phenylalanine hydroxylase-stimulating protein PHS pt Pterin carbinolamine dehydratase Pterin-4-alpha-carbinolamine dehydratase TCF1; 6-pyruvoyl-tetrahydropterin synthase/dimerization cofactor of hepatocyte nuclear factor 1 alpha (TCF1); DCoH; Dimerization cofactor of hepatocyte nuclear factor 1-alpha; dimerization cofactor of HNF1; dimerizing cofactor for HNF1; PCD; Phenylalanine hydroxylase-stimulating protein; PHS; Pterin carbinolamine dehydratase; pterin-4 alpha-carbinolamine dehydratase/dimerization cofactor of hepatocyte nuclear factor 1 alpha; Pterin-4-alpha-carbinolamine dehydratase
Gene Aliases: DCOH; PCBD; PCBD1; PCD; PHS; TCF1
UniProt ID: (Human) P61457, (Mouse) P61458, (Rat) P61459
Entrez Gene ID: (Human) 5092, (Mouse) 13180, (Rat) 29700
Molecular Function:
dehydratase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.