Proteintech
Manufacturer Code:164701AP150UL
Catalog # 50-173-0966
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PC6 Fusion Protein Ag9552 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hPC6 PC5 PC6 PC6A PCSK5 Proprotein convertase 5 Proprotein convertase 6 SPC6; PC5; PC6; prohormone convertase 5; Proprotein convertase 5; Proprotein convertase 6; proprotein convertase PC5; Proprotein convertase subtilisin/kexin type 5; protease PC6; rPC5; SPC6; subtilase; Subtilisin-like proprotein convertase 6; Subtilisin/kexin-like protease PC5
Gene Aliases: b2b1549Clo; b2b585Clo; PC5; PC5/6A; PC5A; PC6; PC6A; PCSK5; SPC6
UniProt ID: (Human) Q92824, (Rat) P41413, (Mouse) Q04592
Entrez Gene ID: (Human) 5125, (Rat) 116548, (Mouse) 18552
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.