Invitrogen
Manufacturer Code:PA540482
Catalog # PIPA540482
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human PBX3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Homeobox protein PBX3; pre B cell leukemia homeobox 3; pre B-cell leukemia transcription factor 3; pre-B-cell leukemia homeobox 3; Pre-B-cell leukemia transcription factor 3
Gene Aliases: PBX3
UniProt ID: (Human) P40426, (Mouse) O35317
Entrez Gene ID: (Human) 5090, (Bovine) 539222, (Rat) 311876, (Guinea pig) 100729729, (Mouse) 18516
Molecular Function: DNA-directed RNA polymerase RNA binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.