Proteintech
Manufacturer Code:125711AP150UL
Catalog # 50-172-8077
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PBX3 Fusion Protein Ag3239 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Homeobox protein PBX3; Homeobox protein PBX3 PBX3 pre B cell leukemia homeobox 3; pre-B-cell leukemia homeobox 3; Pre-B-cell leukemia transcription factor 3
Gene Aliases: PBX3
UniProt ID: (Human) P40426, (Mouse) O35317
Entrez Gene ID: (Human) 5090, (Rat) 311876, (Mouse) 18516
Molecular Function: DNA-directed RNA polymerase RNA binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.