OriGene
Manufacturer Code:UM500077CF
Catalog # 50-168-4980
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | UMAB110 |
Immunogen | Full length human recombit protein of human PBX1 produced in HEK293 cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CAKUHED; Homeobox protein PBX1; Homeobox protein PRL; pre-B-cell leukemia homeobox 1; Pre-B-cell leukemia transcription factor 1
Gene Aliases: PBX1; PRL
UniProt ID: (Human) P40424
Entrez Gene ID: (Human) 5087
Molecular Function:
DNA-directed RNA polymerase
RNA binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.