Invitrogen
Manufacturer Code:MA537814
Catalog # PIMA537814
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1820 |
Immunogen | A synthesized peptide derived from human PBRM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 2310032M22Rik; 2310032M22Rik 2610016F04Rik AI507524 BAF180 BRG1-associated factor 180 hPB1 PB1 PBRM1 polybromo 1 Polybromo-1D Protein polybromo-1 RGD1565549; BAF180; BRG1-associated factor 180; hPB1; polybromo 1; Polybromo-1D; Protein polybromo-1
Gene Aliases: 2610016F04Rik; AI507524; BAF180; PB1; PBRM1; RGD1565549
UniProt ID: (Human) Q86U86
Entrez Gene ID: (Human) 55193, (Rat) 306254, (Mouse) 66923
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.