Proteintech
Manufacturer Code:125631AP150UL
Catalog # 50-172-8070
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PBRM1 Fusion Protein Ag3148 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310032M22Rik; BAF180; BAF180 BRG1 associated factor 180 hPB1 PB1 PBRM1 polybromo 1 Polybromo 1D Protein polybromo 1; BRG1-associated factor 180; hPB1; polybromo 1; Polybromo-1D; Protein polybromo-1
Gene Aliases: 2610016F04Rik; AI507524; BAF180; PB1; PBRM1; RGD1565549
UniProt ID: (Human) Q86U86
Entrez Gene ID: (Human) 55193, (Rat) 306254, (Mouse) 66923
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.