Novus Biologicals
Manufacturer Code:NBP1457690.025MG
Catalog # NB018582
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide surrounding amino acid 81 of mouse PBR (BAA04749). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: benzodiazepine peripheral binding site; benzodiazepine peripheral binding site BZRPDBI IBP MBRPBS mDRC Mitochondrial benzodiazepine receptor PBRBPBS PBRS peripheral benzodiazepine receptor Peripheral-type benzodiazepine receptor pk18 PKBSbenzodiazapine receptor (peripheral) PTBR translocator protein translocator protein (18kDa); Mitochondrial benzodiazepine receptor; PBR; Peripheral-type benzodiazepine receptor; PKBS; Translocator protein; translocator protein (18kDa)
Gene Aliases: BPBS; BZRP; DBI; IBP; MBR; mDRC; PBR; PBS; pk18; PKBS; PTBR; TSPO
UniProt ID: (Human) P30536
Entrez Gene ID: (Human) 706
Molecular Function: receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.