Novus Biologicals
Manufacturer Code:NBP273262AF405
Catalog # N273262A405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7G5 |
Immunogen | Full length recombinant protein of human PBLD (NP_071412) produced in HEK293T cells. |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EC 5.1 FLJ14767 FLJ35507 MAWBPMAWDBPMAWD binding protein MAWD-binding protein phenazine biosynthesis-like domain-containing protein phenazine biosynthesis-like protein domain containing Unknown protein 32 from 2D-page of liver tissue; epididymis secretory protein Li 306; MAWD-binding protein; MAWDBP; Phenazine biosynthesis-like domain-containing protein; phenazine biosynthesis-like protein domain containing; Unknown protein 32 from 2D-page of liver tissue
Gene Aliases: HEL-S-306; MAWBP; MAWDBP; PBLD
UniProt ID: (Human) P30039
Entrez Gene ID: (Human) 64081
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.