OriGene
Manufacturer Code:TA501994
Catalog # 50-167-3089
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7G5 |
Immunogen | Full length human recombit protein of human PBLD produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory protein Li 306; HEL-S-306 MAWBP MAWDBP; MAWD-binding protein; MAWDBP; Phenazine biosynthesis-like domain-containing protein; phenazine biosynthesis-like protein domain containing; Unknown protein 32 from 2D-page of liver tissue
Gene Aliases: HEL-S-306; MAWBP; MAWDBP; PBLD
UniProt ID: (Human) P30039
Entrez Gene ID: (Human) 64081
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.