Invitrogen
Manufacturer Code:PA558220
Catalog # PIPA558220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PBLD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory protein Li 306; MAWD-binding protein; MAWDBP; Phenazine biosynthesis-like domain-containing protein; phenazine biosynthesis-like protein domain containing; Unknown protein 32 from 2D-page of liver tissue
Gene Aliases: HEL-S-306; MAWBP; MAWDBP; PBLD
UniProt ID: (Human) P30039
Entrez Gene ID: (Human) 64081
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.