Invitrogen
Manufacturer Code:PA544517
Catalog # PIPA544517
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human PBLD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory protein Li 306; MAWD binding protein; MAWD binding protein homolog 1; MAWD-binding protein; MAWDBP; Phenazine biosynthesis-like domain-containing protein; Phenazine biosynthesis-like domain-containing protein 1; phenazine biosynthesis-like protein domain containing; sphenazine biosynthesis-like protein domain containing; Unknown protein 32 from 2D-page of liver tissue
Gene Aliases: 0610038K03Rik; HEL-S-306; MAWBP; Mawbp2; MAWDBP; PBLD; Pbld1
UniProt ID: (Bovine) Q2HJF4, (Human) P30039, (Rat) Q68G31, (Mouse) Q9DCG6
Entrez Gene ID: (Rabbit) 100342477, (Pig) 100737003, (Dog) 479226, (Bovine) 504417, (Horse) 100063067, (Human) 64081, (Guinea pig) 100717573, (Rat) 171564, (Mouse) 68371
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.