Invitrogen
Manufacturer Code:PA5112642
Catalog # PIPA5112642
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Phenazine biosynthesis-like domain-containing protein (11-288AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610038K03Rik BOS_24183 epididymis secretory protein Li 306 HEL-S-306 LOC100342477 MAWBP Mawbp2 MAWD binding protein MAWD binding protein homolog 1 MAWD-binding protein MAWDBP PBLD Pbld1 phenazine biosynthesis like protein domain containing Phenazine biosynthesis-like domain-containing protein Phenazine biosynthesis-like domain-containing protein 1 phenazine biosynthesis-like protein domain containing phenazine biosynthesis-like protein domain containing 1 sphenazine biosynthesis-like protein domain containing Unknown protein 32 from 2D-page of liver tissue; epididymis secretory protein Li 306; MAWD binding protein homolog 1; MAWD-binding protein; MAWDBP; Phenazine biosynthesis-like domain-containing protein; Phenazine biosynthesis-like domain-containing protein 1; phenazine biosynthesis-like protein domain containing; Unknown protein 32 from 2D-page of liver tissue
Gene Aliases: 0610038K03Rik; HEL-S-306; MAWBP; Mawbp2; MAWDBP; PBLD; Pbld1
UniProt ID: (Human) P30039, (Mouse) Q9DCG6
Entrez Gene ID: (Human) 64081, (Mouse) 68371
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.