Novus Biologicals
Manufacturer Code:NBP299521G
Catalog # NB083620
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant PBK (NP_060962.2 Met 1-Val 322). |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cancer/testis antigen 84; Cancer/testis antigen 84 CT84MAPKK-like protein kinase EC 2.7.12.2 FLJ14385 Nori-3PDZ-binding kinase PDZ binding kinase serine/threonine protein kinase Spermatogenesis-related protein kinase SPKT-LAK cell-originated protein kinase TOPKlymphokine-activated killer T-cell-originated protein kinase; CT84; epididymis luminal protein 164; Lymphokine-activated killer T-cell-originated protein kinase; MAPKK-like protein kinase; Nori-3; PDZ-binding kinase; serine/threonine protein kinase; Spermatogenesis-related protein kinase; SPK; T-LAK cell-originated protein kinase
Gene Aliases: CT84; HEL164; Nori-3; PBK; SPK; TOPK
UniProt ID: (Human) Q96KB5
Entrez Gene ID: (Human) 55872
Molecular Function: kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.