R&D Systems
Manufacturer Code:MAB43351100
Catalog # MAB43351100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 882108 |
Immunogen | <em>E. coli</em>-derived recombinant human PBEF/Visfatin Pro27-His491 Accession # P43490 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
Protein Aliases: EC 2.4.2.12 MGC117256 NAmPRTase NAMPT nicotinamide phosphoribosyltransferase NMPRTase PBEF1 PBEF1110035O14Rik PBEF1DKFZp666B131 Pre-B cell-enhancing factor pre-B-cell colony enhancing factor 1 Pre-B-cell colony-enhancing factor 1 VF VISFATIN; NAmPRTase; Nicotinamide phosphoribosyltransferase; Pre-B cell-enhancing factor; pre-B-cell colony enhancing factor 1; Pre-B-cell colony-enhancing factor 1; Visfatin
Gene Aliases: 1110035O14Rik; NAMPT; PBEF; PBEF1; VF; VISFATIN
UniProt ID: (Human) P43490
Entrez Gene ID: (Human) 10135
Molecular Function:
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.