Invitrogen
Manufacturer Code:MA1117
Catalog # PIMA1117
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1F8-3A8 |
Immunogen | Purified internal fragment of human recombit PAX8 expressed in E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: paired box 8 paired box homeotic gene 8 Paired Box Protein Pax-8 Paired Domain Gene 8; paired box gene 8; Paired box protein Pax-8; paired domain gene 8
Gene Aliases: Pax-8; PAX8
UniProt ID: (Human) Q06710, (Mouse) Q00288
Entrez Gene ID: (Human) 7849, (Mouse) 18510
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.