Invitrogen
Manufacturer Code:MA516389
Catalog # PIMA516389
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SP34 |
Immunogen | Synthetic peptide from C-terminus of human Pax-5 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: B-cell lineage specific activator; B-cell-specific transcription factor; BSAP; BSAP B cell specific activator protein B-cell lineage specific activator B-cell-specific transcription factor paired box homeotic gene 5 paired box protein Pax-5 transcription factor PAX 5; paired box gene 5 (B-cell lineage specific activator); paired box homeotic gene 5; Paired box protein Pax-5; paired domain gene 5; transcription factor PAX 5
Gene Aliases: ALL3; BSAP; PAX5; RGD1565236
UniProt ID: (Human) Q02548
Entrez Gene ID: (Dog) 612021, (Human) 5079, (Rat) 500453
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.