Invitrogen
Manufacturer Code:PA572630
Catalog # PIPA572630
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 258-284 amino acids from the Central region of DANRE pax2a |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: No isthmus protein; paired box gene 2.1; paired box gene 2a; paired box homeotic gene 2a; Paired box protein Pax-2a; Paired box protein Pax-2a No isthmus protein Pax&lsqbZf-b] pax2a noi pax21 paxzf-b; Pax[Zf-b]
Gene Aliases: cb378; noi; Pax-2; pax-b; pax2.1; pax2a; pax2a1; pax[zf-b]; paxb; PAXZF-B
UniProt ID: (Zebrafish) Q90268
Entrez Gene ID: (Zebrafish) 30425
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.