Abnova
Manufacturer Code:PAB18776
Catalog # 89-120-332
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 15 amino acids at internal region of human PAWR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of PAWR.
For Research Use Only
Protein Aliases: Par-4; PAR4; PRKC apoptosis WT1 regulator protein; PRKC, apoptosis, WT1, regulator; Prostate apoptosis response 4 protein; prostate apoptosis response protein 4; prostate apoptosis response protein PAR-4; prostate apoptosis response-4; transcriptional repressor PAR4; WT1-interacting protein
Gene Aliases: Par-4; PAR4; PAWR
UniProt ID: (Human) Q96IZ0
Entrez Gene ID: (Human) 5074
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.